missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, puriss. p.a., ACS Reagent, ≥99%, Solstice
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -80°C |
| Color | Colorless |
| Density | 0.801g/mL at 25°C |
| Boiling Point | 117°C |
| Flash Point | 14°C (57.2°F) |
| Odor | Characteristic |
| Packaging | Glass Bottle |
| Linear Formula | (CH3)2CHCH2COCH3 |
| Quantity | 2.5 L |
| UN Number | UN1245 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard2' | P210-P261-P305 + P351 + P338 |
| DOTInformation | Class 3 - PG 2 - Methyl isobutyl ketone |
| RTECSNumber | 203-550-1 |