missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone Oxime 98.0+%, TCI America™

Catalog No. p-7132279
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M064225ML 25 mL
M0642500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M064225ML Supplier TCI America Supplier No. M064225ML

Chemical Identifiers

CAS 105-44-2
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00013933
InChI Key ZKALVNREMFLWAN-SREVYHEPSA-N
Synonym 2-pentanone, 4-methyl-, oxime, methyl isobutyl oxime, methyl isobutyl ketoxime, usaf am-4, 2-methyl-4-pentanone oxime, 4-methyl-2-pentanone oxime, 4-methylpentan-2-one oxime, unii-nzo785t1sg, nzo785t1sg, 2e-4-methyl-2-pentanone oxime #
PubChem CID 5354076
IUPAC Name (Z)-N-(4-methylpentan-2-ylidene)hydroxylamine
SMILES CC(C)C\C(C)=N/O

Specifications

Color Yellow
Boiling Point 70°C
Quantity 25 mL
Formula Weight 115.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone Oxime
EINECSNumber (2)-0546
RTECSNumber SA9450000
TSCA Yes