missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-Pentanone, OmniSolv™ MilliporeSigma™

Catalog No. MMX13011
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX13011 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX13011 Supplier Technidata Supplier No. MX13011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For HPLC, Spectrophotometry and Gas Chromatography

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point -83.9°C
Color Colorless to Yellow
Flash Point 17.85°C (64.1°F)
Odor Minty
Packaging Glass Bottle
Quantity 4 L
UN Number UN 1245
Solubility Information Water: Partially soluble
Formula Weight 100.16
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-Pentanone
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 200-714-4
Recommended Storage Room Temperature; Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.