missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, HPLC Grade, 99+%

Catalog No. AA43170M6
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43170M6 4 x 4 L
AA43170K2 1 L
AA43170M1 4 x 1 L
AA43170K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA43170M6 Supplier Thermo Scientific Chemicals Supplier No. 043170M6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%, Color (APHA) 10 | CAS: 108-10-1 | C6H12O | 100.161 g/mol

4-Methyl-2-pentanone is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point °C to 84°C
Color Colorless
Density 0.801
Boiling Point 117°C to 118°C
Flash Point 13°C (55°F)
Ignition Point 460°C (860°F)
Odor Mild
Assay Percent Range >99%
Refractive Index 1.396
Quantity 4 x 4 L
Beilstein 605399
Solubility Information Miscible with ethanol,ether,acetone,benzene and chloroform. Slightly miscible with water.
Vapor Pressure 21 hPa (16mm Hg) at 20°C
Formula Weight 100.16
Grade HPLC
Concentration or Composition (by Analyte or Components) 4-Methyl-2-pentanone
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone, Filtered through 0.2μ filters.
Show More Show Less
Hazard Category H225-H319-H332-H335-H351
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-550-1
RTECSNumber SA9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only