missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, CHROMASOLV™, for HPLC, ≥99.5%, Solstice

Catalog No. 6000990
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000990 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000990 Supplier Solstice Supplier No. 2932611L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™, for HPLC,≥99.5%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Absorbance max. 0.01 at 400 nm, max. 0.02 at 380 nm, max. 0.15 at 360 nm, max. 0.50 at 340 nm, max. 1.0 at 335 nm
CAS Max % 100.0000%
Melting Point -80°C (literature)
Density 0.801 g/mL at 25 °C(lit.)
Boiling Point 115°C to 117°C
Flash Point 14°C
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2COCH3
Quantity 1 L
UN Number UN1245
Beilstein 605399
Formula Weight 100.16
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material 4-Methyl-2-pentanone, CHROMASOLV™, for HPLC, ≥99.5%
Show More Show Less
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methyl isobutyl ketone
EINECSNumber 203-550-1
Recommended Storage Room Temp