missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, ACS, 98.5%, Spectrum™ Chemical

Catalog No. p-7165874
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608276 100 mL Amber Glass Bottle
18608277 500 mL Amber Glass Bottle
18608271 1 L Amber Glass Bottle
18608272 4 L Amber Glass Bottle
18608273 4 L Poly-Coated Glass Bottle
18608275 20 L Steel Can
18608274 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18608276 Supplier Spectrum Chemical Mfg Cor Supplier No. M1275100ML
Only null left

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(C)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 mL
Residue after Evaporation 5e-05
Percent Purity 0.985
Grade ACS
Titratable Acid 0.002 meq/g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.