missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, ACS, 98.5%, Spectrum™ Chemical

Catalog No. 18608277
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608277 500 mL Amber Glass Bottle
18608276 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608277 Supplier Spectrum Chemical Mfg Cor Supplier No. M1275500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(C)=O

Specifications

CAS Max % 1
Boiling Point 114°C to 118°C (237.2°F to 244.4°F)
Packaging Amber Glass Bottle
Quantity 500 mL
Residue after Evaporation 5e-05
Percent Purity 0.985
Grade ACS
Titratable Acid 0.002 meq/g
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.