missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, ACS, 98.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 114°C to 118°C (237.2°F to 244.4°F) |
| Packaging | Amber Glass Bottle |
| Quantity | 100 mL |
| Residue after Evaporation | 5e-05 |
| Percent Purity | 0.985 |
| Grade | ACS |
| Titratable Acid | 0.002 meq/g |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Methyl-2-pentanone |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.