missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, ACS, 98.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Drum |
| Quantity | 200 L |
| Residue after Evaporation | 5e-05 |
| Percent Purity | 0.985 |
| Grade | ACS |
| Titratable Acid | 0.002 meq/g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.