missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, 99.5%, spectrophotometric grade

CAS: 108-10-1 | C6H12O | 100.16 g/mol

Supplier:  Thermo Scientific Chemicals 222170025

 View more versions of this product

Catalog No. AC222170025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methyl-2-pentanone
108-10-1
0.01 at 400nm, 0.02 at 380nm, 0.15 at 360nm, 0.5 at 340nm, 1 at 335nm
-84.0°C
117.4°C
99.5% min. (GC)
C6H12O
(CH3)2CHCH2COCH3
2.5 L
0.001% max.
15,5253
Solubility in water: 17g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,methylene chloride and acetone
CC(C)CC(=O)C
100.16
CHEBI:82344
99.5%
99.5%
99.4
100.0
0.8000g/mL
14°C
Glass bottle
1.3950 to 1.3970
MFCD00008938
01,691
0.8
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methylpentan-2-one
7909
100.16
Liquid

Chemical Identifiers

108-10-1
100.16
NTIZESTWPVYFNL-UHFFFAOYSA-N
7909
4-methylpentan-2-one
C6H12O
MFCD00008938
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CHEBI:82344
CC(C)CC(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-550-1

RTECSNumber : SA9275000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only