missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 97.0+%, TCI America™
$74.56 - $249.78
Chemical Identifiers
| CAS | 635305-48-5 |
|---|---|
| Molecular Formula | C11H17BO2S |
| Molecular Weight (g/mol) | 224.13 |
| MDL Number | MFCD11045430 |
| InChI Key | FNRMPXJVCSMVMA-UHFFFAOYSA-N |
| Synonym | 4,4,5,5-Tetramethyl-2-(4-methyl-2-thienyl)-1,3,2-dioxaborolane, 4-Methyl-2-thiopheneboronic Acid Pinacol Ester |
| PubChem CID | 10443592 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-(4-methylthiophen-2-yl)-1,3,2-dioxaborolane |
| SMILES | CC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1 |
Chemical Identifiers
| 635305-48-5 | |
| 224.13 | |
| FNRMPXJVCSMVMA-UHFFFAOYSA-N | |
| 10443592 | |
| CC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1 |
| C11H17BO2S | |
| MFCD11045430 | |
| 4,4,5,5-Tetramethyl-2-(4-methyl-2-thienyl)-1,3,2-dioxaborolane, 4-Methyl-2-thiopheneboronic Acid Pinacol Ester | |
| 4,4,5,5-tetramethyl-2-(4-methylthiophen-2-yl)-1,3,2-dioxaborolane |
Specifications
| 635305-48-5 | |
| White-Yellow | |
| C11H17BO2S | |
| 1 g | |
| FNRMPXJVCSMVMA-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-(4-methylthiophen-2-yl)-1,3,2-dioxaborolane | |
| 10443592 | |
| ≥97.0% (GC) | |
| 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene |
| 31°C | |
| 93°C | |
| MFCD11045430 | |
| 4,4,5,5-Tetramethyl-2-(4-methyl-2-thienyl)-1,3,2-dioxaborolane, 4-Methyl-2-thiopheneboronic Acid Pinacol Ester | |
| CC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1 | |
| 224.13 | |
| 224.13 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator