missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(2-methyl-1-propenyl)tetrahydropyran (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. M236325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M236325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M236325G Supplier TCI America Supplier No. M236325G
Only null left

Chemical Identifiers

CAS 16409-43-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00036607
InChI Key CZCBTSFUTPZVKJ-UHFFFAOYSA-N
PubChem CID 27866
IUPAC Name 4-methyl-2-(2-methylprop-1-enyl)oxane
SMILES CC1CCOC(C1)C=C(C)C

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 g
Formula Weight 154.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-(2-methyl-1-propenyl)tetrahydropyran (cis- and trans- mixture)
EINECSNumber (5)-0680
RTECSNumber UQ1470000
TSCA Yes