missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-[1-(tert-butoxycarbonyl)piperid-4-yl]-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC31101CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC31101CB 250mg Amber Glass Bottle
CC31101DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31101CB Supplier Thermo Scientific Chemicals Supplier No. CC31101CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 216955-61-2
Molecular Formula C15H22N2O4S
Molecular Weight (g/mol) 326.41
MDL Number MFCD06659070
InChI Key XDKKXEMKXSGKOR-UHFFFAOYSA-N
Synonym 4-methyl-2-1-tert-butoxycarbonyl piperid-4-yl-1,3-thiazole-5-carboxylic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-4-methylthiazole-5-carboxylic acid, 2-1-tert-butoxycarbonyl-4-piperidinyl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-5-carboxy-4-methyl-1,3-thiazole, tert-butyl 4-5-carboxy-4-methyl-1,3-thiazol-2-yl piperidin-1-carboxylate, 4-methyl-2-1-2-methylpropan-2-yl oxycarbonyl piperidin-4-yl-1,3-thiazole-5-carboxylic acid
PubChem CID 2795533
IUPAC Name 4-methyl-2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C1CCN(CC1)C(=O)OC(C)(C)C)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 326.41
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-[1-(tert-butoxycarbonyl)piperid-4-yl]-1, 3-thiazole-5-carboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.