missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1H-pyrazole, 97+%

Catalog No. AAA1808306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1808306 5 g
AAA1808303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1808306 Supplier Thermo Scientific Chemicals Supplier No. A1808306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7554-65-6 | C4H6N2 | 82.106 g/mol

4-Methyl-1H-pyrazole is used as an antidote in methanol and ethylene glycol poisoning. It is also employed as an alcohol dehydrogenase inhibitor. It acts a ligand in coordination chemistry. Further, it serves as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-1H-pyrazole is used as an antidote in methanol and ethylene glycol poisoning. It is also employed as an alcohol dehydrogenase inhibitor. It acts a ligand in coordination chemistry. Further, it serves as an intermediate in organic synthesis.

Solubility
Miscible with water, ether, benzene and alcohol.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents, strong acids, acid chlorides, acid anhydrides and chloroformates.

Chemical Identifiers

CAS 7554-65-6
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.106
MDL Number MFCD00005245
InChI Key RIKMMFOAQPJVMX-UHFFFAOYSA-N
Synonym 4-methylpyrazole, fomepizole, antizol, 1h-pyrazole, 4-methyl, fomepizol, 4-methylpyrazol, fomepizolum, fomepizol inn-spanish, fomepizolum inn-latin, fomepizole usan:inn
PubChem CID 3406
ChEBI CHEBI:5141
IUPAC Name 4-methyl-1H-pyrazole
SMILES CC1=CNN=C1

Specifications

Melting Point 13°C
Density 1.018
Boiling Point 204°C to 207°C
Flash Point 96°C (204°F)
Refractive Index 1.4945
Quantity 5 g
Beilstein 105204
Sensitivity Air Sensitive
Merck Index 14,4216
Solubility Information Miscible with water,ether,benzene and alcohol.
Formula Weight 82.11
Percent Purity ≥97%
Chemical Name or Material 4-Methyl-1H-pyrazole
Show More Show Less
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-445-0
RTECSNumber UQ7350000
TSCA No

RUO – Research Use Only