missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentyne 98.0+%, TCI America™

Catalog No. p-7132084
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02691ML 1 mL
M02695ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M02691ML Supplier TCI America Supplier No. M02691ML
Only null left

Chemical Identifiers

CAS 7154-75-8
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00039857
InChI Key OXRWICUICBZVAE-UHFFFAOYSA-N
Synonym 4-methyl-1-pentyne, 1-pentyne, 4-methyl, isobutylacetylene, isobutylethyne, 4-methylpentine, 4-methylpentyne, 4-methyl pentyne, isobutyl acetylene, 4-methyl-pentyne, 4-methyl-pent-1-yne
PubChem CID 138948
IUPAC Name 4-methylpent-1-yne
SMILES CC(C)CC#C

Specifications

Color Colorless
Boiling Point 61°C
Quantity 1 mL
UN Number 3295
Formula Weight 82.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-pentyne