missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentene 97.0+%, TCI America™

Catalog No. M039225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M039225ML 25 mL
M0392500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M039225ML Supplier TCI America Supplier No. M039225ML

Chemical Identifiers

CAS 691-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008949
InChI Key WSSSPWUEQFSQQG-UHFFFAOYSA-N
Synonym 4-methyl-1-pentene, 1-pentene, 4-methyl, poly 4-methyl-1-pentene, unii-x10hrj2y7w, x10hrj2y7w, poly 4-methylpentene, isobutylethene, isohexene, 4-methylpentene-1, 1-pentene, 4-methyl-, homopolymer
PubChem CID 12724
IUPAC Name 4-methylpent-1-ene
SMILES CC(C)CC=C

Specifications

Melting Point -153°C
Color Colorless
Boiling Point 53°C
Quantity 25 mL
UN Number 2288
Formula Weight 84.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-pentene
EINECSNumber (2)-0031
TSCA Yes