missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentene, 98+%

Catalog No. AA3302909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3302909 10 g
AA3302918 50 g
AA3302930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3302909 Supplier Thermo Scientific Chemicals Supplier No. 03302909

CAS: 691-37-2 | C6H12 | 84.16 g/mol

4-Methyl-1-pentene is used as a monomer for olefin polymerisation. The resulting polymer is poly(4-methyl-1-pentene). It can also be used to produce 1,2-diiodo-4-methyl-pentane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 691-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008949
InChI Key WSSSPWUEQFSQQG-UHFFFAOYSA-N
Synonym 4-methyl-1-pentene, 1-pentene, 4-methyl, poly 4-methyl-1-pentene, unii-x10hrj2y7w, x10hrj2y7w, poly 4-methylpentene, isobutylethene, isohexene, 4-methylpentene-1, 1-pentene, 4-methyl-, homopolymer
PubChem CID 12724
IUPAC Name 4-methylpent-1-ene
SMILES CC(C)CC=C

Specifications

Melting Point -153°C
Color Colorless
Density 0.664 g/mL
Boiling Point 53°C to 54°C
Flash Point -32°C (-25°F)
Odor Petrol-like
Refractive Index 1.382
Linear Formula (CH3)2CHCH2CH=CH2
Quantity 10 g
UN Number UN3295
Beilstein 1731096
Solubility Information Soluble in alcohol,benzene,chloroform,petroleum ether. Insoluble in water.
Formula Weight 84.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-pentene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 211-720-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only