missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentanol 99.0+%, TCI America™

Catalog No. p-7132355
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M077510ML 10 mL
M077525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M077510ML Supplier TCI America Supplier No. M077510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-89-1
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002962
InChI Key PCWGTDULNUVNBN-UHFFFAOYSA-N
Synonym 4-methyl-1-pentanol, isohexanol, isohexyl alcohol, 4-methylpentanol, iso-hexanol, 2-methyl-5-pentanol, 1-pentanol, 4-methyl, pentanol, 4-methyl, 4-methyl-pentan-1-ol, unii-x796xfp7d4
PubChem CID 12296
ChEBI CHEBI:63910
IUPAC Name 4-methylpentan-1-ol
SMILES CC(C)CCCO

Specifications

Color Colorless
Boiling Point 152°C
Quantity 10 mL
UN Number 1987
Formula Weight 102.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-pentanol
EINECSNumber (2)-0217
RTECSNumber NR3020000
TSCA No