missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-cyclohexene, TCI America™

Catalog No. p-7132052
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M020125ML 25 mL
M0201500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M020125ML Supplier TCI America Supplier No. M020125ML
Only null left

Chemical Identifiers

CAS 591-47-9
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00001573
InChI Key FSWCCQWDVGZMRD-UHFFFAOYSA-N
PubChem CID 11572
IUPAC Name 4-methylcyclohexene
SMILES CC1CCC=CC1

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 mL
UN Number 3295
Formula Weight 96.17
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-cyclohexene