missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7132714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14125G 5 g
M141225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14125G Supplier TCI America Supplier No. M14125G
Only null left

Chemical Identifiers

CAS 34885-03-5
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00152628,MFCD27997499,MFCD31705071
InChI Key OSINZLLLLCUKJH-UHFFFAOYSA-N
Synonym 1-(Hydroxymethyl)-4-methylcyclohexane
PubChem CID 118193
IUPAC Name (4-methylcyclohexyl)methanol
SMILES CC1CCC(CO)CC1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture)