missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-oxazole-5-carboxylic acid, 95%, Thermo Scientific™

Catalog No. p-4510008
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC10001CB 250 mg
CC10001DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC10001CB Supplier Thermo Scientific Chemicals Supplier No. CC10001CB

Chemical Identifiers

CAS 2510-32-9
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
InChI Key ZIXUNDOOBLSXPE-UHFFFAOYSA-N
Synonym 4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl
PubChem CID 292311
IUPAC Name 4-methyl-1,3-oxazole-5-carboxylic acid
SMILES CC1=C(OC=N1)C(=O)O

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 127.1
Percent Purity 95%
Chemical Name or Material 4-Methyl-1, 3-oxazole-5-carboxylic acid