missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-dioxolane 98.0+%, TCI America™

Catalog No. m091625ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M091625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M091625ML Supplier TCI America Supplier No. M091625ML

Chemical Identifiers

CAS 1072-47-5
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00014109
InChI Key SBUOHGKIOVRDKY-UHFFFAOYSA-N
Synonym 1,3-dioxolane, 4-methyl, 1,3-dioxolane,4-methyl, acmc-1bvod, 4-methyl-1,3-dioxolan-2-yl, 4-methyl-1,3-dioxolane
PubChem CID 66119
IUPAC Name 4-methyl-1,3-dioxolane
SMILES CC1COCO1

Specifications

Color Colorless
Boiling Point 85°C
Quantity 25 mL
UN Number 1993
Formula Weight 88.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1,3-dioxolane