missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3,2-dioxathiolane 2-Oxide (mixture of isomers) 98.0+%, TCI America™

Catalog No. p-7133237
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
M24715G 5 g
M247125G 25 g
2 options

Catalog No. M24715G

Supplier: TCI America M24715G

Only null left

Chemical Identifiers

CAS 1469-73-4
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.138
MDL Number MFCD00041209
InChI Key SJHAYVFVKRXMKG-UHFFFAOYSA-N
Synonym 1,2-Propanediol Cyclic Sulfite, 1,2-Propyleneglycol Sulfite, 1,2-Propylene Sulfite
PubChem CID 15117
IUPAC Name 4-methyl-1,3,2-dioxathiolane 2-oxide
SMILES CC1COS(=O)O1

Specifications

Color Yellow
Boiling Point 40°C
Quantity 5 g
Formula Weight 122.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-1,3,2-dioxathiolane 2-Oxide (mixture of isomers)
RTECSNumber WT3500000
TSCA No