missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2,3-thiadiazole-5-carboxylic acid, 98%

Catalog No. p-7025114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2044906 5 g
AAB2044914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2044906 Supplier Thermo Scientific Chemicals Supplier No. B2044906
Only null left

CAS: 18212-21-0 | C4H3N2O2S | 143.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18212-21-0
Molecular Formula C4H3N2O2S
Molecular Weight (g/mol) 143.14
MDL Number MFCD00173830
InChI Key NHHQOYLPBUYHQU-UHFFFAOYSA-M
Synonym 4-methyl-1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl, pubchem10092, acmc-209xfo, 4-methyl-1,2,3-thiadiazole-5-carboxylicacid, tos-bb-0050, 4-methyl-1,2,3 thiadiazol-5-carboxylic acid, 4-methyl-5-hydroxycarbonyl-1,2,3-thiadiazole, 4methyl-1,2,3 thiadiazole-5-carboxylic acid
PubChem CID 1381992
IUPAC Name 4-methylthiadiazole-5-carboxylic acid
SMILES CC1=C(SN=N1)C([O-])=O

Specifications

Melting Point 184°C to 186°C
Quantity 5 g
Formula Weight 144.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-1,2,3-thiadiazole-5-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only