missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride, Thermo Scientific™

Catalog No. p-4510607
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC02302DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC02302DA Supplier Thermo Scientific Chemicals Supplier No. CC02302DA

Chemical Identifiers

CAS 59944-65-9
Molecular Formula C4H3ClN2OS
Molecular Weight (g/mol) 162.59
MDL Number MFCD00173831
InChI Key SDNDOCTUXWLDIX-UHFFFAOYSA-N
Synonym 4-methyl-1,2,3-thiadiazole-5-carbonyl chloride, 4-methyl-1,2,3-thiadiazole-5-carbonylchloride, 1,2,3-thiadiazole-5-carbonylchloride, 4-methyl, 4-methyl-1,2,3 thiadiazole-5-carbonyl chloride, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid chloride, 4-methyl-5-thiadiazolecarbonyl chloride, 5-chlorocarbonyl-4-methyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-5-carbonyl chloride, 4-methyl-9ci
PubChem CID 2736982
SMILES CC1=C(SN=N1)C(Cl)=O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 4-methylthiadiazole-5-carbonyl chloride
Formula Weight 162.6
Percent Purity 90%
Chemical Name or Material 4-Methyl-1, 2, 3-thiadiazole-5-carbonyl chloride