missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,10-phenanthroline

Catalog No. p-7049950
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3188803 1 g
AA3188806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3188803 Supplier Thermo Scientific Chemicals Supplier No. 03188803

CAS: 31301-28-7 | C13H10N2 | 194.237 g/mol

4-methyl-1,10-phenanthroline can be used to produce 1,10-Phenanthrolin-4-aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31301-28-7
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00004978
InChI Key NAZZKEZTSOOCSZ-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 4-methyl, 4-methylpyridino 3,2-h quinoline, acmc-1aihj, 4-methyl-1,10 phenanthroline, 4-methyl-1,10-phenanthroline, 4-methyl-1,10-phenanthroline1,10-phenanthroline, 4-methyl
PubChem CID 93149
IUPAC Name 4-methyl-1,10-phenanthroline
SMILES CC1=C2C=CC3=C(C2=NC=C1)N=CC=C3

Specifications

Melting Point 144°C to 145°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 194.24
Physical Form Crystalline
Chemical Name or Material 4-Methyl-1,10-phenanthroline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only