missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxypyridine 98.0+%, TCI America™

Catalog No. p-7132814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16965G 5 g
M169625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16965G Supplier TCI America Supplier No. M16965G
Only null left

Chemical Identifiers

CAS 620-08-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
InChI Key XQABVLBGNWBWIV-UHFFFAOYSA-N
Synonym pyridine, 4-methoxy, methyl pyridin-4-yl ether, 4-methoxy pyridine, .gamma.-methoxypyridine, 4-methoxypyridin, 4-methoxypyrdine, 4-methoxylpyridine, 4-methoxy-pyridine, 4-methoxypyridine, pubchem2585
PubChem CID 69278
IUPAC Name 4-methoxypyridine
SMILES COC1=CC=NC=C1

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 190°C
Quantity 5 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methoxypyridine