missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxypyridine N-Oxide 98.0+%, TCI America™
$136.08 - $136.08
Chemical Identifiers
| CAS | 1122-96-9 |
|---|---|
| Molecular Formula | C6H7NO2 |
| Molecular Weight (g/mol) | 125.127 |
| MDL Number | MFCD00006207 |
| InChI Key | BOFAIBPJCWFJFT-UHFFFAOYSA-N |
| Synonym | 4-methoxypyridine n-oxide, 4-methoxypyridine 1-oxide, 4-methoxypyridine-n-oxide, pyridine, 4-methoxy-, 1-oxide, 4-methoxypyridin-1-ium-1-olate, 4-methoxypyridine-n-oxide hydrate, pubchem2587, acmc-1buuj, 4-methoxypyridinen-oxide, 4-methoxypiridine-n-oxide |
| PubChem CID | 70743 |
| IUPAC Name | 4-methoxy-1-oxidopyridin-1-ium |
| SMILES | COC1=CC=[N+](C=C1)[O-] |
Chemical Identifiers
| 1122-96-9 | |
| 125.127 | |
| BOFAIBPJCWFJFT-UHFFFAOYSA-N | |
| 70743 | |
| COC1=CC=[N+](C=C1)[O-] |
| C6H7NO2 | |
| MFCD00006207 | |
| 4-methoxypyridine n-oxide, 4-methoxypyridine 1-oxide, 4-methoxypyridine-n-oxide, pyridine, 4-methoxy-, 1-oxide, 4-methoxypyridin-1-ium-1-olate, 4-methoxypyridine-n-oxide hydrate, pubchem2587, acmc-1buuj, 4-methoxypyridinen-oxide, 4-methoxypiridine-n-oxide | |
| 4-methoxy-1-oxidopyridin-1-ium |
Specifications
| 1122-96-9 | |
| C6H7NO2 | |
| 5 g | |
| BOFAIBPJCWFJFT-UHFFFAOYSA-N | |
| 4-methoxy-1-oxidopyridin-1-ium | |
| 70743 | |
| ≥98.0% (GC,T) | |
| 4-Methoxypyridine N-Oxide |
| 112°C | |
| MFCD00006207 | |
| 4-methoxypyridine n-oxide, 4-methoxypyridine 1-oxide, 4-methoxypyridine-n-oxide, pyridine, 4-methoxy-, 1-oxide, 4-methoxypyridin-1-ium-1-olate, 4-methoxypyridine-n-oxide hydrate, pubchem2587, acmc-1buuj, 4-methoxypyridinen-oxide, 4-methoxypiridine-n-oxide | |
| COC1=CC=[N+](C=C1)[O-] | |
| 125.127 | |
| 125.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No