missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxypyridine, 98+%

Catalog No. p-7047683
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1962506 5 g
AAL1962514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1962506 Supplier Thermo Scientific Chemicals Supplier No. L1962506

CAS: 620-08-6 | C6H7NO | 109.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 620-08-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00674049
InChI Key XQABVLBGNWBWIV-UHFFFAOYSA-N
Synonym pyridine, 4-methoxy, methyl pyridin-4-yl ether, 4-methoxy pyridine, .gamma.-methoxypyridine, 4-methoxypyridin, 4-methoxypyrdine, 4-methoxylpyridine, 4-methoxy-pyridine, 4-methoxypyridine, pubchem2585
PubChem CID 69278
IUPAC Name 4-methoxypyridine
SMILES COC1=CC=NC=C1

Specifications

Melting Point 4°C
Density 1.075
Boiling Point 191°C
Flash Point 74°C (165°F)
Refractive Index 1.521
Quantity 5 g
Beilstein 108211
Formula Weight 109.13
Percent Purity ≥98%
Chemical Name or Material 4-Methoxypyridine
Hazard Category H227-H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-624-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only