missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylmagnesium bromide, 0.5M in THF

Catalog No. p-7052384
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA89435AE 100 mL
AA89435AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA89435AE Supplier Thermo Scientific Chemicals Supplier No. 089435AE

CAS: 13139-86-1 | C7H7BrMgO | 211.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13139-86-1
Molecular Formula C7H7BrMgO
Molecular Weight (g/mol) 211.34
MDL Number MFCD00013515
InChI Key JKXAZEXIOPFJSC-UHFFFAOYSA-M
Synonym 4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent
PubChem CID 2734898
SMILES COC1=CC=C([Mg]Br)C=C1

Specifications

Color Brown
Density 0.955 g/mL
Flash Point -20°C (-4°F)
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
IUPAC Name bromo(4-methoxyphenyl)magnesium
Formula Weight 211.34
Concentration or Composition (by Analyte or Components) 0.5M in THF
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylmagnesium bromide
Show More Show Less
Hazard Category H225-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only