missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl beta-D-Glucopyranoside 97.0+%, TCI America™
$350.44 - $1049.46
Chemical Identifiers
| CAS | 6032-32-2 |
|---|---|
| Molecular Formula | C13H18O7 |
| Molecular Weight (g/mol) | 286.28 |
| MDL Number | MFCD06797143 |
| InChI Key | SIXFVXJMCGPTRB-UJPOAAIJSA-N |
| Synonym | methylarbutin, 4-methoxyphenyl beta-d-glucopyranoside, p-methoxyphenyl b-d-glucoside, unii-f9mw5fu0xh, 4-methoxyphenyl b-d-glucopyranoside, f9mw5fu0xh, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-4-methoxyphenoxy oxane-3,4,5-triol, methylarbutoside, 4-methoxyphenylglucoside, p-methoxyphenyl beta-d-glucopyranoside |
| PubChem CID | 80131 |
| ChEBI | CHEBI:81207 |
| IUPAC Name | (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol |
| SMILES | COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O |
Chemical Identifiers
| 6032-32-2 | |
| 286.28 | |
| SIXFVXJMCGPTRB-UJPOAAIJSA-N | |
| 80131 | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol |
| C13H18O7 | |
| MFCD06797143 | |
| methylarbutin, 4-methoxyphenyl beta-d-glucopyranoside, p-methoxyphenyl b-d-glucoside, unii-f9mw5fu0xh, 4-methoxyphenyl b-d-glucopyranoside, f9mw5fu0xh, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-4-methoxyphenoxy oxane-3,4,5-triol, methylarbutoside, 4-methoxyphenylglucoside, p-methoxyphenyl beta-d-glucopyranoside | |
| CHEBI:81207 | |
| COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O |
Specifications
| 6032-32-2 | |
| White | |
| MFCD06797143 | |
| methylarbutin, 4-methoxyphenyl beta-d-glucopyranoside, p-methoxyphenyl b-d-glucoside, unii-f9mw5fu0xh, 4-methoxyphenyl b-d-glucopyranoside, f9mw5fu0xh, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-4-methoxyphenoxy oxane-3,4,5-triol, methylarbutoside, 4-methoxyphenylglucoside, p-methoxyphenyl beta-d-glucopyranoside | |
| COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O | |
| 286.28 | |
| CHEBI:81207 | |
| ≥97.0% (HPLC) | |
| 4-Methoxyphenyl beta-D-Glucopyranoside |
| 176°C | |
| C13H18O7 | |
| 5 g | |
| SIXFVXJMCGPTRB-UJPOAAIJSA-N | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol | |
| 80131 | |
| 286.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer