missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. p-7130746
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08255G 5 g
H082525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08255G Supplier TCI America Supplier No. H08255G

Chemical Identifiers

CAS 50687-62-2
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00191515
InChI Key MTQSVJUCLQYISS-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid 4-Methoxyphenyl Ester, 4-Methoxyphenylparaben
PubChem CID 578384
IUPAC Name (4-methoxyphenyl) 4-hydroxybenzoate
SMILES COC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)O

Specifications

Melting Point 194°C
Color White
Quantity 5 g
Formula Weight 244.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 4-Hydroxybenzoate