missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 3-O-Benzyl-4,6-O-benzylidene-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. M16401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16401G 1 g
M16405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16401G Supplier TCI America Supplier No. M16401G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 303127-81-3
Molecular Formula C27H28O7
Molecular Weight (g/mol) 464.51
MDL Number MFCD08276394
InChI Key JQCLKKJUWALUTI-UHFFFAOYNA-N
PubChem CID 126970120
IUPAC Name 8-(benzyloxy)-6-(4-methoxyphenoxy)-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-ol
SMILES COC1=CC=C(OC2OC3COC(OC3C(OCC3=CC=CC=C3)C2O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 207°C
Color White
Quantity 1 g
Formula Weight 464.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 3-O-Benzyl-4,6-O-benzylidene-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer