missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 3-O-Allyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-beta-D-glucopyranoside 97.0+%, TCI America™

Catalog No. M15981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15981G 1 g
M15985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M15981G Supplier TCI America Supplier No. M15981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 889453-84-3
Molecular Formula C31H29NO8
Molecular Weight (g/mol) 543.572
InChI Key KDSWOYVINHSEOI-YLVCXTSJSA-N
PubChem CID 44629794
IUPAC Name 2-[(4aR,6S,7R,8R,8aS)-6-(4-methoxyphenoxy)-2-phenyl-8-prop-2-enoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]isoindole-1,3-dione
SMILES COC1=CC=C(C=C1)OC2C(C(C3C(O2)COC(O3)C4=CC=CC=C4)OCC=C)N5C(=O)C6=CC=CC=C6C5=O

Specifications

Melting Point 130°C
Color White
Quantity 1 g
Formula Weight 543.57
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 3-O-Allyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-beta-D-glucopyranoside
TSCA No
Recommended Storage Refrigerator