missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 3-O-Allyl-2-azido-4,6-O-benzylidene-2-deoxy-beta-D-galactopyranoside 95.0+%, TCI America™

Catalog No. m16431g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16431G Supplier TCI America Supplier No. M16431G

Chemical Identifiers

CAS 889453-83-2
Molecular Formula C23H25N3O6
Molecular Weight (g/mol) 439.468
InChI Key TYEQNJAZZVSANI-YYKQMJRKSA-N
PubChem CID 91659134
IUPAC Name (4aR,6S,7R,8R,8aR)-7-azido-6-(4-methoxyphenoxy)-2-phenyl-8-prop-2-enoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine
SMILES COC1=CC=C(C=C1)OC2C(C(C3C(O2)COC(O3)C4=CC=CC=C4)OCC=C)N=[N+]=[N-]

Specifications

Color White
Quantity 1 g
Formula Weight 439.47
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 3-O-Allyl-2-azido-4,6-O-benzylidene-2-deoxy-beta-D-galactopyranoside
TSCA No
Recommended Storage Freezer