missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 2-O-Acetyl-3-O-allyl-4,6-O-benzylidene-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. M20651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20651G 1 g
M20655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20651G Supplier TCI America Supplier No. M20651G

Chemical Identifiers

CAS 1477956-18-5
Molecular Formula C25H28O8
Molecular Weight (g/mol) 456.491
InChI Key AYSHRBCEMWANHR-HGGAONJVSA-N
PubChem CID 91659153
IUPAC Name [(4aR,6S,7R,8S,8aR)-6-(4-methoxyphenoxy)-2-phenyl-8-prop-2-enoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl] acetate
SMILES CC(=O)OC1C(C2C(COC(O2)C3=CC=CC=C3)OC1OC4=CC=C(C=C4)OC)OCC=C

Specifications

Color White
Quantity 1 g
Formula Weight 456.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2-O-Acetyl-3-O-allyl-4,6-O-benzylidene-beta-D-glucopyranoside
TSCA No
Recommended Storage Refrigerator