missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl 2,4,6-Tri-O-acetyl-3-O-benzyl-beta-D-glucopyranoside 96.0+%, TCI America™
$248.72 - $748.05
Chemical Identifiers
| CAS | 303127-79-9 |
|---|---|
| Molecular Formula | C26H30O10 |
| Molecular Weight (g/mol) | 502.516 |
| InChI Key | MDZXNXKMNWQSDZ-WSGIOKLISA-N |
| PubChem CID | 10839164 |
| IUPAC Name | [(2R,3R,4S,5R,6S)-3,5-diacetyloxy-6-(4-methoxyphenoxy)-4-phenylmethoxyoxan-2-yl]methyl acetate |
| SMILES | CC(=O)OCC1C(C(C(C(O1)OC2=CC=C(C=C2)OC)OC(=O)C)OCC3=CC=CC=C3)OC(=O)C |
Chemical Identifiers
| 303127-79-9 | |
| 502.516 | |
| 10839164 | |
| CC(=O)OCC1C(C(C(C(O1)OC2=CC=C(C=C2)OC)OC(=O)C)OCC3=CC=CC=C3)OC(=O)C |
| C26H30O10 | |
| MDZXNXKMNWQSDZ-WSGIOKLISA-N | |
| [(2R,3R,4S,5R,6S)-3,5-diacetyloxy-6-(4-methoxyphenoxy)-4-phenylmethoxyoxan-2-yl]methyl acetate |
Specifications
| 303127-79-9 | |
| White | |
| 1 g | |
| CC(=O)OCC1C(C(C(C(O1)OC2=CC=C(C=C2)OC)OC(=O)C)OCC3=CC=CC=C3)OC(=O)C | |
| 502.516 | |
| 502.52 | |
| Crystalline Powder |
| 160°C | |
| C26H30O10 | |
| MDZXNXKMNWQSDZ-WSGIOKLISA-N | |
| [(2R,3R,4S,5R,6S)-3,5-diacetyloxy-6-(4-methoxyphenoxy)-4-phenylmethoxyoxan-2-yl]methyl acetate | |
| 10839164 | |
| ≥96.0% (HPLC) | |
| 4-Methoxyphenyl 2,4,6-Tri-O-acetyl-3-O-benzyl-beta-D-glucopyranoside |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator