missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxymethyl)piperidine hydrochloride

Catalog No. p-7033842
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
AAH51020MD 250 mg
AAH5102003 1 g
2 options

Catalog No. AAH51020MD

Supplier: Thermo Scientific Chemicals H51020MD

Only null left

CAS: 916317-00-5 | C7H16ClNO | 165.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 916317-00-5
Molecular Formula C7H16ClNO
Molecular Weight (g/mol) 165.66
MDL Number MFCD08062602
InChI Key FLRDPZJPRQWKLD-UHFFFAOYSA-N
Synonym 4-methoxymethyl piperidine hydrochloride, 4-methoxymethyl piperidine hcl, 4-methoxymethyl-piperidine hydrochloride, 4-methoxymethylpiperidine hydrochloride, 4-methoxymethyl-piperidinehydrochloride, acmc-20a6g3, methoxymethylpiperidinehydrochloride, methoxy-4-piperidylmethane, chloride, 4-methoxymethyl piperidinehcl, 4-methoxymethyl piperidinehydrochloride
PubChem CID 17218599
IUPAC Name 4-(methoxymethyl)piperidine;hydrochloride
SMILES Cl.COCC1CCNCC1

Specifications

Quantity 250 mg
Formula Weight 165.66
Chemical Name or Material 4-(Methoxymethyl)piperidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only