missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyindole 98.0+%, TCI America™

Catalog No. p-7132619
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12501G 1 g
M12505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12501G Supplier TCI America Supplier No. M12501G
Only null left

Chemical Identifiers

CAS 4837-90-5
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00009737
InChI Key LUNOXNMCFPFPMO-UHFFFAOYSA-N
Synonym 4-methoxyindole, 1h-indole, 4-methoxy, 4-methoxylindole, 1h-indol-4-yl methyl ether, indole, 4-methoxy, 4-methoxy-indole, zlchem 710, pubchem7433, acmc-1aoxe
PubChem CID 138363
IUPAC Name 4-methoxy-1H-indole
SMILES COC1=CC=CC2=C1C=CN2

Specifications

Melting Point 70°C
Boiling Point 181°C
Quantity 1 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyindole