missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyindole-2-carboxylic acid, 97+%
CAS: 103260-65-7 | C10H9NO3 | 191.186 g/mol
Supplier: Thermo Scientific Chemicals H5239206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Methoxyindole-2-carboxylic acid | |
| C10H9NO3 | |
| 5 g | |
| ZZAVIQXQBBOHBB-UHFFFAOYSA-N | |
| 4-methoxy-1H-indole-2-carboxylic acid | |
| 907248 | |
| ≥97% |
| 103260-65-7 | |
| MFCD02664458 | |
| 4-methoxyindole-2-carboxylic acid, 4-methoxy-indole-2-carboxylic acid, 4-methoxyindole-2carboxylic acid, 1h-indole-2-carboxylicacid, 4-methoxy, 1h-indole-2-carboxylic acid, 4-methoxy, 4-methoxy-indole-2-carboxylicacid, pubchem9380, acmc-20aa7f, 4-methoxy-2-indolecarboxylic acid | |
| COC1=CC=CC2=C1C=C(N2)C(=O)O | |
| 191.186 | |
| 191.19 |
Chemical Identifiers
| 103260-65-7 | |
| 191.186 | |
| ZZAVIQXQBBOHBB-UHFFFAOYSA-N | |
| 907248 | |
| COC1=CC=CC2=C1C=C(N2)C(=O)O |
| C10H9NO3 | |
| MFCD02664458 | |
| 4-methoxyindole-2-carboxylic acid, 4-methoxy-indole-2-carboxylic acid, 4-methoxyindole-2carboxylic acid, 1h-indole-2-carboxylicacid, 4-methoxy, 1h-indole-2-carboxylic acid, 4-methoxy, 4-methoxy-indole-2-carboxylicacid, pubchem9380, acmc-20aa7f, 4-methoxy-2-indolecarboxylic acid | |
| 4-methoxy-1H-indole-2-carboxylic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only