missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyindole-2-carboxylic acid, 97+%

Catalog No. AAH52392MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52392MD 250 mg
AAH5239203 1 g
AAH5239206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52392MD Supplier Thermo Scientific Chemicals Supplier No. H52392MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103260-65-7 | C10H9NO3 | 191.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103260-65-7
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD02664458
InChI Key ZZAVIQXQBBOHBB-UHFFFAOYSA-N
Synonym 4-methoxyindole-2-carboxylic acid, 4-methoxy-indole-2-carboxylic acid, 4-methoxyindole-2carboxylic acid, 1h-indole-2-carboxylicacid, 4-methoxy, 1h-indole-2-carboxylic acid, 4-methoxy, 4-methoxy-indole-2-carboxylicacid, pubchem9380, acmc-20aa7f, 4-methoxy-2-indolecarboxylic acid
PubChem CID 907248
IUPAC Name 4-methoxy-1H-indole-2-carboxylic acid
SMILES COC1=CC=CC2=C1C=C(N2)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 191.19
Percent Purity ≥97%
Chemical Name or Material 4-Methoxyindole-2-carboxylic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only