missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxyformanilide 98.0+%, TCI America™

Catalog No. M16235G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16235G Supplier TCI America Supplier No. M16235G

Chemical Identifiers

CAS 5470-34-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00086774
InChI Key SXEVZVMJNXOXIJ-UHFFFAOYSA-N
Synonym N-Formyl-4-methoxyaniline, N-(4-Methoxyphenyl)formamide
PubChem CID 79616
IUPAC Name N-(4-methoxyphenyl)formamide
SMILES COC1=CC=C(C=C1)NC=O

Specifications

Melting Point 80°C
Boiling Point 159°C
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Methoxyformanilide