missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxycoumarin, 98%

Catalog No. AAA1367014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1367014 25 g
AAA1367006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1367014 Supplier Thermo Scientific Chemicals Supplier No. A1367014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20280-81-3 | C10H8O3 | 176.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20280-81-3
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00017349
InChI Key MLCMXDYMSAZNPC-UHFFFAOYSA-N
PubChem CID 529426
IUPAC Name 4-methoxychromen-2-one
SMILES COC1=CC(=O)OC2=CC=CC=C21

Specifications

Melting Point 122°C to 125°C
Quantity 25 g
Beilstein 145960
Synonym 4-methoxycoumarin, 4-methoxycourmarin, 4-methoxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 4-methoxy, 2h-1-benzopyran-2-one,4-methoxy, 4-methoxy-2h-chromen-2-on, 4-methoxycoumarine, acmc-20amex, coumarin, 4-methoxy
Formula Weight 176.17
Percent Purity 98%
Chemical Name or Material 4-Methoxycoumarin
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only