missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxycinnamic Acid 98.0+%, TCI America™
Supplier: TCI America M0576100G
Specifications
| 4-Methoxycinnamic Acid | |
| 190°C | |
| C10H10O3 | |
| 100 g | |
| AFDXODALSZRGIH-QPJJXVBHSA-N | |
| (E)-3-(4-methoxyphenyl)prop-2-enoic acid | |
| 699414 | |
| ≥98.0% (GC,T) |
| 830-09-1 | |
| White | |
| MFCD00004398 | |
| 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid | |
| COC1=CC=C(C=C1)C=CC(=O)O | |
| 178.187 | |
| 178.19 | |
| Crystalline Powder |
Chemical Identifiers
| 830-09-1 | |
| 178.187 | |
| AFDXODALSZRGIH-QPJJXVBHSA-N | |
| 699414 | |
| COC1=CC=C(C=C1)C=CC(=O)O |
| C10H10O3 | |
| MFCD00004398 | |
| 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid | |
| (E)-3-(4-methoxyphenyl)prop-2-enoic acid |
Safety and Handling
EINECSNumber : (3)-3927
RTECSNumber : UD3391300
TSCA : Yes