missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxycinnamic Acid 98.0+%, TCI America™

Catalog No. M057625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M057625G 25 g
M0576100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M057625G Supplier TCI America Supplier No. M057625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 830-09-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00004398
InChI Key AFDXODALSZRGIH-QPJJXVBHSA-N
Synonym 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid
PubChem CID 699414
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 190°C
Color White
Quantity 25 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxycinnamic Acid
EINECSNumber (3)-3927
RTECSNumber UD3391300
TSCA Yes