Learn More
4-Methoxycinnamic acid, 99%
CAS: 830-09-1 | C10H10O3 | 178.187 g/mol
Supplier: Thermo Scientific Chemicals H6014414
Description
It exerts anti-hyperglycemic/ hypoglycemic effect by stimulating insulin secretion from pancreas. It could be developed into a new potential therapeutic agent used in type 2 diabetic patients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt exerts anti-hyperglycemic/ hypoglycemic effect by stimulating insulin secretion from pancreas. It could be developed into a new potential therapeutic agent used in type 2 diabetic patients.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool dry conditions. Keep away from oxidizing agents.
Specifications
| 4-Methoxycinnamic acid | |
| 830-09-1 | |
| C10H10O3 | |
| 25 g | |
| Soluble in dimethyl sulfoxide and methanol. | |
| COC1=CC=C(C=C1)C=CC(=O)O | |
| 178.187 | |
| 178.18 |
| predominantly trans | |
| 170°C to 173°C | |
| MFCD00004398 | |
| 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid | |
| AFDXODALSZRGIH-QPJJXVBHSA-N | |
| (E)-3-(4-methoxyphenyl)prop-2-enoic acid | |
| 699414 | |
| 99% |
Chemical Identifiers
| 830-09-1 | |
| 178.187 | |
| AFDXODALSZRGIH-QPJJXVBHSA-N | |
| 699414 | |
| COC1=CC=C(C=C1)C=CC(=O)O |
| C10H10O3 | |
| MFCD00004398 | |
| 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid | |
| (E)-3-(4-methoxyphenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-594-0
RTECSNumber : UD3391300
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only