missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxychalcone, 97%

Catalog No. p-7026251
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2215806 5 g
AAB2215814 25 g
AAB2215822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2215806 Supplier Thermo Scientific Chemicals Supplier No. B2215806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 959-23-9 | C16H14O2 | 238.286 g/mol

4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

Solubility
Soluble in dichloromethane and methanol. Insoluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.

Chemical Identifiers

CAS 959-23-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00008407
InChI Key KJHHAPASNNVTSN-KPKJPENVSA-N
Synonym 4'-methoxychalcone, 2e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, 1-4-methoxyphenyl-3-phenylprop-2-en-1-one, chembl34398, 1-4-methoxyphenyl-3-phenyl-2-propen-1-one, e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, unii-bm45n45fiz, e-1-4-methoxy-phenyl-3-phenyl-propenone, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl-, 2e, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl
PubChem CID 641818
IUPAC Name (E)-1-(4-methoxyphenyl)-3-phenylprop-2-en-1-one
SMILES COC1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2

Specifications

Melting Point 101°C to 103°C
Quantity 5 g
Beilstein 644640
Solubility Information Soluble in dichloromethane and methanol. Insoluble in water.
Formula Weight 238.29
Percent Purity 97%
Chemical Name or Material 4'-Methoxychalcone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-495-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only