missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$46.20 - $61.61
Chemical Identifiers
| CAS | 99768-12-4 |
|---|---|
| Molecular Formula | C8H9BO4 |
| MDL Number | MFCD01632203 |
| InChI Key | PQCXFUXRTRESBD-UHFFFAOYSA-N |
| PubChem CID | 2734369 |
Specifications
| 99768-12-4 | |
| White-Yellow | |
| MFCD01632203 | |
| methyl 4-boronobenzoate, 4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl phenyl boronic acid, 4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonylphenyl boronic acid, methyl4-boronobenzoate, 4-methoxycarbonylbenzeneboronic acid, p-methoxycarbonyl phenylboronic acid, 4-methoxycabonyl phenylboronic acid | |
| B(C1=CC=C(C=C1)C(=O)OC)(O)O | |
| 179.966 | |
| 179.97 | |
| 4-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride) |
| 234°C | |
| C8H9BO4 | |
| 1 g | |
| PQCXFUXRTRESBD-UHFFFAOYSA-N | |
| (4-methoxycarbonylphenyl)boronic acid | |
| 2734369 | |
| Crystalline Powder |
Safety and Handling
TSCA : No