missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M19071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19071G 1 g
M19075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19071G Supplier TCI America Supplier No. M19071G

Chemical Identifiers

CAS 99768-12-4
Molecular Formula C8H9BO4
MDL Number MFCD01632203
InChI Key PQCXFUXRTRESBD-UHFFFAOYSA-N
PubChem CID 2734369

Specifications

Melting Point 234°C
Color White-Yellow
Quantity 1 g
Synonym methyl 4-boronobenzoate, 4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl phenyl boronic acid, 4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonylphenyl boronic acid, methyl4-boronobenzoate, 4-methoxycarbonylbenzeneboronic acid, p-methoxycarbonyl phenylboronic acid, 4-methoxycabonyl phenylboronic acid
SMILES B(C1=CC=C(C=C1)C(=O)OC)(O)O
IUPAC Name (4-methoxycarbonylphenyl)boronic acid
Molecular Weight (g/mol) 179.966
Formula Weight 179.97
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)