missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)bicyclo[2.2.2]octan-1-carboxylic Acid 98.0+%, TCI America™

Catalog No. m28551g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M28551G Supplier TCI America Supplier No. M28551G

Specifications

CAS 18720-35-9
Melting Point 188°C
Color White
Molecular Formula C11H16O4
Quantity 1 g
MDL Number MFCD18459359
Synonym Monomethyl Hydrogen Bicyclo[2.2.2]octane-1,4-dicarboxylate, Bicyclo[2.2.2]octane-1,4-dicarboxylic Acid Monomethyl Ester
InChI Key KBZVAQVEHVGIEI-UHFFFAOYSA-N
SMILES COC(=O)C12CCC(CC1)(CC2)C(O)=O
IUPAC Name 4-(methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid
Molecular Weight (g/mol) 212.25
PubChem CID 11042062
Formula Weight 212.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methoxycarbonyl)bicyclo[2.2.2]octan-1-carboxylic Acid
Show More Show Less